Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532099
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532099 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532099 Supplier Invitrogen™ Supplier No. PA532099
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: HCT116. Predicted reactivity: Mouse (84%), Rat (82%), Bovine (85%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a member of the phospholipase C family, which catalyze the hydrolysis of phosphatidylinositol 4, 5-bisphosphate to generate the second messengers diacylglycerol and inositol 1, 4, 5-trisphosphate (IP3). Diacylglycerol and IP3 mediate a variety of cellular responses to extracellular stimuli by inducing protein kinase C and increasing cytosolic Ca(2+) concentrations. This enzyme localizes to the plasma membrane and requires calcium for activation. Its activity is inhibited by spermine, sphingosine, and several phospholipids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCD3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene PLCD3
Gene Accession No. Q8N3E9
Gene Alias 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-3; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-3; 2610205J15Rik; Kiaa1964; mKIAA1964; phosphoinositide phospholipase C-delta-3; phospholipase C delta 3; phospholipase C, delta 3; phospholipase C-delta-3; PLC delta3; Plcd3; PLC-delta-3
Gene Symbols PLCD3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PLCD3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 113026
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.