Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLCD3 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA547717
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
This gene encodes a member of the phospholipase C family, which catalyze the hydrolysis of phosphatidylinositol 4, 5-bisphosphate to generate the second messengers diacylglycerol and inositol 1, 4, 5-trisphosphate (IP3). Diacylglycerol and IP3 mediate a variety of cellular responses to extracellular stimuli by inducing protein kinase C and increasing cytosolic Ca(2+) concentrations. This enzyme localizes to the plasma membrane and requires calcium for activation. Its activity is inhibited by spermine, sphingosine, and several phospholipids.
Specifications
| PLCD3 | |
| Polyclonal | |
| Unconjugated | |
| PLCD3 | |
| 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-3; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-3; 2610205J15Rik; Kiaa1964; mKIAA1964; phosphoinositide phospholipase C-delta-3; phospholipase C delta 3; phospholipase C, delta 3; phospholipase C-delta-3; PLC delta3; Plcd3; PLC-delta-3 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 113026, 72469 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| Q8K2J0, Q8N3E9 | |
| PLCD3 | |
| E. coli-derived recombinant human PLC-delta3 Lys148-Ala344. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction