Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCD4 Polyclonal Antibody

Catalog No. PIPA5121678
Change view
Click to view available options
Quantity:
20 μL
200 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121678 20 μL
PIPA5119344 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. PIPA5121678 Supplier Invitrogen™ Supplier No. PA5121678
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PLCD4 Polyclonal Antibody for Western Blot, IHC (P)

This gene encodes a member of the delta class of phospholipase C enzymes. Phospholipase C enzymes play a critical role in many cellular processes by hydrolyzing phosphatidylinositol 4,5-bisphosphate into two intracellular second messengers, inositol 1,4,5-trisphosphate and diacylglycerol. Expression of this gene may be a marker for cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCD4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4
Gene Plcd4
Gene Accession No. Q8K3R3, Q9BRC7
Gene Alias 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase delta-4; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase delta-4; 4921507K24Rik; hPLCD4; phosphoinositide phospholipase C delta-4; phosphoinositide phospholipase C-delta-4; phospholipase C delta 4; phospholipase C, delta 4; phospholipase C-delta-4; PLC delta4; Plcd; PLCD4; PLC-delta-4
Gene Symbols Plcd4
Host Species Rabbit
Immunogen Recombinant protein Phospholipase C Delta 4. The antigen corresponds to amino acid range 1-250 of the target protein.
Purification Method Antigen affinity chromatography, Protein A
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18802, 84812
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.