Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCE1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553333
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553333 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553333 Supplier Invitrogen™ Supplier No. PA553333
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGISPLGNQS VIIETGRAHP DSRRAVFHFH YEVDRRMSDT FCTLSENLIL DDCGNCVPLP GGEEKQKKNY VAYTCKLMEL AKNCDNKNEQ LQCDHCDTLN DKYFCFEGSC EKVDMVYSGD SFCRKDFTDS QAAKT Highest antigen sequence identity to the following orthologs: Mouse - 59%, Rat - 57%.

This gene encodes a phospholipase enzyme that catalyzes the hydrolysis of phosphatidylinositol-4,5-bisphosphate to generate two second messengers: inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG). These second messengers subsequently regulate various processes affecting cell growth, differentiation, and gene expression. This enzyme is regulated by small monomeric GTPases of the Ras and Rho families and by heterotrimeric G proteins. In addition to its phospholipase C catalytic activity, this enzyme has an N-terminal domain with guanine nucleotide exchange (GEF) activity. Mutations in this gene cause early-onset nephrotic syndrome; characterized by proteinuria, edema, and diffuse mesangial sclerosis or focal and segmental glomerulosclerosis. Alternative splicing results in multiple transcript variants encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCE1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLCE1
Gene Accession No. Q9P212
Gene Alias 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase epsilon-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase epsilon-1; 4933403A21Rik; KIAA1516; mKIAA1516; NPHS3; pancreas-enriched phospholipase C; phosphoinositide phospholipase C; phosphoinositide phospholipase C-epsilon-1; phosphoinositide-specific phospholipase C epsilon-1; phospholipase C epsilon 1; phospholipase C, epsilon 1; phospholipase C-epsilon-1; PLCE; PLCE1; PLCepsilon; PLC-epsilon-1; PPLC
Gene Symbols PLCE1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLCE1. Recombinant protein control fragment (Product #RP-90791).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51196
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.