Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCG1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596768
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596768 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596768 Supplier Invitrogen™ Supplier No. PA596768
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: MCF7, Jurkat, HepG2, Mouse brain, Mouse lung, Rat brain; Cellular Location: Cell projection, lamellipodium, ruffle Immunogen sequence: NSYAISFRAE GKIKHCRVQQ EGQTVMLGNS EFDSLVDLIS YYEKHPLYRK MKLRYPINEE ALEKIGTAEP DYGALYEGRN PGFYVEANPM PTFKCAVKAL F.

The mammalian Phospholipase Cg (PLCg) family has two closely related proteins, PLCg1 and PLCg2. The PLCg isozymes have the core structure domains and a unique array of domains containing an additional PH domain, two SH2 domains and one SH3 domain. In response to extracellular stimuli, such as hormones and growth factors, receptor tyrosine kinases (RTKs) phosphorylate and activate PLCg. Activated PLCg catalyzes hydrolysis of phosphatidylinositol 4,5-bisphosphate (PIP2) to produce the metabolic second messenger molecules inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCG1
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.38 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PLCG1
Gene Accession No. P10686, P19174, Q62077
Gene Alias 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma 1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1; AI894140; Cded; cell differentiation and embryonic development; EC 3.1.4.11; inositoltrisphosphohydrolase; monophosphatidylinositol phosphodiesterase; NCKAP3; OTTHUMP00000178982; phosphatidylinositol phospholipase C; phosphoinositidase C; phosphoinositide phospholipase C; phosphoinositide phospholipase C-gamma-1; phospholipase C gamma 1; phospholipase C, gamma 1; phospholipase C, gamma 1 (formerly subtype 148); phospholipase C-1; phospholipase C-148; Phospholipase C2; phospholipase C-gamma-1; Phospholipase C-II; PLC gamma1; PLC1; Plc-1; PLC148; PLC-148; PLC2I; Plcg1; Plcg-1; PLCgamma1; Plc-gamma1; PLC-gamma-1; PLC-II; PPLCA; triphosphoinositide phosphodiesterase
Gene Symbols PLCG1
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 700-800 of human PLC gamma 1 (PLCG1) (NP_0026512).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18803, 25738, 5335
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.