Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLCG1, Rabbit anti-Human, Clone: C4, Abnova™

Catalog No. 89335853 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-335-853 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-335-853 Supplier Abnova Corporation Supplier No. MAB19019
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Specifications

Antigen PLCG1
Applications Flow Cytometry
Classification Monoclonal
Clone C4
Conjugate Unconjugated
Dilution Flow Cytometry (1 ug/mL - 0.001 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.02% sodium azide, 0.1% BSA).
Gene phospholipase C, gamma 1
Gene Alias PLC-II/PLC1/PLC148/PLCgamma1
Gene Symbols PLCG1
Host Species Rabbit
Immunogen A synthetic phosphopeptide corresponding to residues surrounding Y783 of human PLCG1.
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5335
Target Species Human
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.