Learn More
Invitrogen™ PLCG2 Polyclonal Antibody

Description
Positive test controls include: U-251MG, Raji, Ramos, HeLa. Immunogen sequence: MSTTVNVDSL AEYEKSQIKR ALELGTVMTV FSFRKSTPER RTVQVIMETR QVAWSKTADK IEGFLDIMEI KEIRPGKNSK DFERAKAVRQ KEDCCFTILY GTQFVLSTLS LAADSKEDAV NWLSGLKILH QEAMNASTPT IIESWLRKQI YSVDQTRRNS ISLRELKTIL PLINFKVSSA KFLKDKFVEI GAHKDELSFE QFHLFYKKLM FEQQKSILDE FKKDSSVFIL GNTDRPDASA VYLHDFQRFL IHEQQEHWAQ DLNKVRERMT KFIDDTMRET AEPFLFVDEF LTYLFSRENS.
Specifications
Specifications
| Antigen | PLCG2 |
| Applications | ELISA, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 |
| Gene | Plcg2 |
| Gene Accession No. | P16885, Q8CIH5 |
| Gene Alias | 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-2; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-2; APLAID; FCAS3; phosphoinositide phospholipase C; phosphoinositide phospholipase C-gamma-2; phospholipase C gamma 2; phospholipase C, gamma 2; phospholipase C, gamma 2 (phosphatidylinositol-specific); phospholipase C-gamma-2; Phospholipase C-IV; PLC gamma 2; PLC gamma2; PLC- gamma2; PLC gamma-2; PLCG2; Plcg-2; PLC-gamma; PLCgamma2; PLC-gamma-2; PLC-IV |
| Gene Symbols | Plcg2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.