Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLCG2, Rabbit anti-Human, FITC, Clone: G3, Abnova™
Rabbit Monoclonal Antibody
Supplier: Abnova Corporation MAB19034
Specifications
PLCG2 | |
Monoclonal | |
FITC | |
In PBS, pH 7.4 (0.2% BSA, 0.09% sodium azide). | |
- | |
Rabbit | |
10 Reactions | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
G3 | |
Flow Cytometry (5 uL/106 cells or 0.05 ug/mL) The optimal working dilution should be determined by the end user. | |
phospholipase C, gamma 2 (phosphatidylinositol-specific) | |
PLCG2 | |
A synthetic phosphopeptide corresponding to residues surrounding Y759 of human PLCG2. | |
RUO | |
5336 | |
Store at 2-8°C. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction