Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLCH1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA566118
Description
Immunogen sequence: ETTKHATNTVY ETTCTPISKT KPDDDLSSKA KTAALESNLP GSPNTSRGWL PKSPTKGEDW ETLKSCSPAS SPDLTLEDVI ADPTLCFNSG E Highest antigen sequence identity to the following orthologs - mouse 54%, rat 54%.
PLCH1 is a member of the PLC-eta family of the phosphoinositide-specific phospholipase C (PLC) superfamily of enzymes that cleave phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2) to generate second messengers inositol 1,4,5-trisphosphate (IP3) and diacylglycerol (DAG).
Specifications
PLCH1 | |
Polyclonal | |
Unconjugated | |
PLCH1 | |
1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase eta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase eta-1; BC042549; Kiaa1069; Phosphoinositide phospholipase C-eta-1; phospholipase C eta 1; phospholipase C, eta 1; phospholipase C-eta-1; phospholipase C-eta1a; phospholipase C-eta1b; phospholipase C-like 3; phospholipase C-like protein 3; PLC eta 1; PLCeta1; PLC-eta-1; Plch1; Plcl3; PLC-L3; RGD1561991 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23007 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q4KWH8 | |
PLCH1 | |
Recombinant Human PLCH1 Isoform B. Recombinant protein control fragment (Product #RP-106808). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction