Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLCH1 Isoform B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA521964
Description
Recommended positive controls: 293T, A431, H1299, HeLaS3, HepG2, Molt-4, Raji. Predicted reactivity: Mouse (99%), Rat (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The production of the second messenger molecules diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3) is mediated by calcium-activated phosphatidylinositol-specific phospholipase C enzymes.
Specifications
PLCH1 Isoform B | |
Polyclonal | |
Unconjugated | |
PLCH1 | |
1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase eta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase eta-1; KIAA1069; Phosphoinositide phospholipase C-eta-1; phospholipase C eta 1; phospholipase C, eta 1; phospholipase C-eta-1; phospholipase C-eta1a; phospholipase C-eta1b; phospholipase C-like 3; phospholipase C-like protein 3; PLC eta 1; PLC-eta-1; PLCH1; PLCL3; PLC-L3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
23007 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Frozen), Western Blot | |
0.93 mg/mL | |
0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7 | |
Q4KWH8 | |
PLCH1 | |
Recombinant fragment corresponding to a region within amino acids 548 and 871 of PLCH1 (Uniprot ID#Q4KWH8). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction