Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCH1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA588853
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA588853 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA588853 Supplier Invitrogen™ Supplier No. PA588853
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LEGNRGKGRA ATSFSLSDVS MLCSDIPDLH STAILQESVI SHLIDNVTLT NENEPGSSIS ALIGQFDETN NQALTVVSHL HNTSVMSGHC PLPSLGLKMP IKHGFCKGKS KSSFLCSSPE LIALSSSETT KHATNTVYET TCTPISKTKP DDDLSSKAKT AALESNLPGS PNTSRGWLPK SPTKGEDWET LKSCSPASSP DLTLEDVIAD PTLCFNSGES SLVEIDGESE NLSLTTCEYR REGTSQLASP LKLKYNQGVV EHFQRGLRNG YCKETLRPSV PEIFNNIQDV KTQSISYLAY Q; Positive Samples: Rat brain; Cellular Location: Cytoplasm, Membrane.

PLCH1 is a member of the PLC-eta family of the phosphoinositide-specific phospholipase C (PLC) superfamily of enzymes that cleave phosphatidylinositol 4,5-bisphosphate (PtdIns(4,5)P2) to generate second messengers inositol 1,4,5-trisphosphate (IP3) and diacylglycerol (DAG).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCH1
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.96 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene PLCH1
Gene Alias 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase eta-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase eta-1; BC042549; Kiaa1069; Phosphoinositide phospholipase C-eta-1; phospholipase C eta 1; phospholipase C, eta 1; phospholipase C-eta-1; phospholipase C-eta1a; phospholipase C-eta1b; phospholipase C-like 3; phospholipase C-like protein 3; PLC eta 1; PLCeta1; PLC-eta-1; Plch1; Plcl3; PLC-L3; RGD1561991
Gene Symbols PLCH1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1100-1400 of human PLCH1 (NP_0558111).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 310463
Target Species Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.