Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLCH2 Polyclonal Antibody
PLCH2 Polyclonal Antibody
Supplier: Thermo Scientific PA584908
Description
PLCH2 Polyclonal Antibody for IHC (P)

Specifications
PLCH2 | |
Polyclonal | |
Unconjugated | |
PLCH2 | |
1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase eta-2; A930027K05Rik; KIAA0450; Phosphoinositide phospholipase C-eta-2; phosphoinositide phospholipase C-like 4; phospholipase C eta 2; phospholipase C, eta 2; phospholipase C-eta2; Phospholipase C-eta-2; phospholipase C-like 4; phospholipase C-like protein 4; PLCeta2; Plc-eta2; PLCH2; PLCL4; PLC-L4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
9651 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75038 | |
PLCH2 | |
Recombinant protein corresponding to Human PLCH2. Recombinant protein control fragment (Product #RP-108606). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction