Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556879
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556879 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556879 Supplier Invitrogen™ Supplier No. PA556879
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LRYLVSRSKQ PLDFMEGNQN TPRFMWLKTV FEAADVDGNG IMLEDTSVEL IKQLNPTLKE AKIRLKFKEI QKSKEKLTTR VTEEEF Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.

Involved in an inositol phospholipid-based intracellular signaling cascade. Shows no PLC activity to phosphatidylinositol 4,5-bisphosphate and phosphatidylinositol. Component in the phospho-dependent endocytosis process of GABA A receptor. Regulates the turnover of receptors and thus contributes to the maintenance of GABA-mediated synaptic inhibition. Its aberrant expression could contribute to the genesis and progression of lung carcinoma. Acts as a inhibitor of PPP1C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLCL1
Gene Accession No. Q15111
Gene Alias 130kDa-Ins(1,4,5)P3 binding protein; C230017K02Rik; inactive phospholipase C-like protein 1; p130; phospholipase C like 1; phospholipase C related, but catalytically inactive protein; phospholipase C, epsilon; phospholipase C, eta 2 pseudogene; phospholipase C-deleted in lung carcinoma; phospholipase C-like 1; phospholipase C-related but catalytically inactive protein; PLCE; PLCL; PLC-L; PLCL1; PLC-L1; PLDL1; PPP1R127; PRIP; PRIP1; PRIP-1; protein phosphatase 1, regulatory subunit 127
Gene Symbols PLCL1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLCL1. Recombinant protein control fragment (Product #RP-96032).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5334
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.