Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCL1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585213
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585213 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585213 Supplier Invitrogen™ Supplier No. PA585213
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Mouse (98%), Rat (99%), Rhesus Monkey (100%), Bovine (98%). Positive Control: Target recombinant protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Involved in an inositol phospholipid-based intracellular signaling cascade. Shows no PLC activity to phosphatidylinositol 4,5-bisphosphate and phosphatidylinositol. Component in the phospho-dependent endocytosis process of GABA A receptor. Regulates the turnover of receptors and thus contributes to the maintenance of GABA-mediated synaptic inhibition. Its aberrant expression could contribute to the genesis and progression of lung carcinoma. Acts as a inhibitor of PPP1C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCL1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.92 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PLCL1
Gene Accession No. Q15111, Q3USB7
Gene Alias 130kDa-Ins(1,4,5)P3 binding protein; C230017K02Rik; inactive phospholipase C-like protein 1; p130; phospholipase C like 1; phospholipase C related, but catalytically inactive protein; phospholipase C, epsilon; phospholipase C, eta 2 pseudogene; phospholipase C-deleted in lung carcinoma; phospholipase C-like 1; phospholipase C-related but catalytically inactive protein; PLCE; PLCL; PLC-L; PLCL1; PLC-L1; PLDL1; PPP1R127; PRIP; PRIP1; PRIP-1; protein phosphatase 1, regulatory subunit 127
Gene Symbols PLCL1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the N-terminus region of human PLCL1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 227120, 5334
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.