Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLCL1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985933
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5933 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5933 Supplier Bioss Supplier No. BS4151R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Involved in an inositol phospholipid-based intracellular signaling cascade. Shows no PLC activity to phosphatidylinositol 4,5-bisphosphate and phosphatidylinositol. Component in the phospho-dependent endocytosis process of GABA A receptor. Regulates the turnover of receptors and thus contributes to the maintenance of GABA-mediated synaptic inhibition. Its aberrant expression could contribute to the genesis and progression of lung carcinoma. Acts as a inhibitor of PPP1C.

Specifications

Antigen PLCL1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene PLCL1
Gene Accession No. Q15111
Gene Alias C230017K02Rik; Inactive phospholipase C-like protein 1; phospholipase C like 1; phospholipase C related, but catalytically inactive protein; phospholipase C, epsilon; phospholipase C, eta 2 pseudogene; Phospholipase C-deleted in lung carcinoma; phospholipase C-like 1; Phospholipase C-related but catalytically inactive protein; PLCE; PLCL; PLC-L; PLCL1; PLC-L1; PLDL1; PPP1R127; PRIP; PRIP-1; protein phosphatase 1, regulatory subunit 127
Gene Symbols PLCL1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human PLCL1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5334
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.