Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLCL2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561557
Description
Immunogen sequence: DMLESSQDNM RTSWVSQMFS EIDVDNLGHI TLCNAVQCIR NLNPGLKTSK IELKFKELHK SKDKAGTEVT KEEFIE Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.
May play an role in the regulation of Ins(1,4,5)P3 around the endoplasmic reticulum.
Specifications
PLCL2 | |
Polyclonal | |
Unconjugated | |
PLCL2 | |
inactive phospholipase C-like protein 2; KIAA1092; mKIAA1092; phospholipase C; phospholipase C like 2; phospholipase C, epsilon 2; phospholipase C-epsilon-2; phospholipase C-L2; phospholipase C-like 2; PLCE2; PLC-epsilon-2; PLC-L(2); Plcl2; PLC-L2; PRIP-2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
23228 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9UPR0 | |
PLCL2 | |
Recombinant protein corresponding to Human PLCL2. Recombinant protein control fragment (Product #RP-96379). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction