Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLCXD1 Antibody (OTI2D7) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273458
Description
PLCXD1 Monoclonal specifically detects PLCXD1 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
PLCXD1 | |
Monoclonal | |
LYOPH | |
Immunohistochemistry 1:150 | |
FLJ11323, phosphatidylinositol-specific phospholipase C, X domain containing 1, PI-PLC X domain-containing protein 1 | |
Mouse | |
36.5 kDa | |
100 μg | |
55344 | |
Human | |
Lyophilized |
Immunohistochemistry | |
OTI2D7 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
PLCXD1 | |
Full length human recombinant protein of human PLCXD1(NP_060860) produced in E.coli. | |
Affinity Purified | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction