Learn More
Invitrogen™ PLCXD1 Monoclonal Antibody (OTI2D7)

Description
PLCXD1 Monoclonal Antibody for IHC (P)
Specifications
Specifications
| Antigen | PLCXD1 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | OTI2D7 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | PLCXD1 |
| Gene Accession No. | Q9NUJ7 |
| Gene Alias | LL0XNC01-136G2.1; phosphatidylinositol specific phospholipase C X domain containing 1; phosphatidylinositol-specific phospholipase C, X domain containing 1; PI-PLC X domain-containing protein 1; PLCXD1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.