Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCXD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA599018
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA599018 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA599018 Supplier Invitrogen™ Supplier No. PA599018
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PLCXD1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene is the most terminal protein-coding gene in the pseudoautosomal (PAR) region on chromosomes X and Y. Alternate splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCXD1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PLCXD1
Gene Accession No. Q9NUJ7
Gene Alias LL0XNC01-136G2.1; phosphatidylinositol specific phospholipase C X domain containing 1; phosphatidylinositol-specific phospholipase C, X domain containing 1; PI-PLC X domain-containing protein 1; PLCXD1
Gene Symbols PLCXD1
Host Species Rabbit
Immunogen Recombinant Human PI-PLC X domain-containing protein 1 protein (1-323AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55344
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.