Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLCXD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563216
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563216 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563216 Supplier Invitrogen™ Supplier No. PA563216
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DFNHFYAMDE THHKCLVLRI QEAFGNKLCP ACSVESLTLR TLWEKNCQVL IFYHCPFYKQ YPFLWPGKKI PAPWANTTS Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 95%.

PLC-XD2 (PI-PLC X domain-containing protein 2) is a 305 amino acid protein that contains a domain that is present in many PLC isozymes, the PI-PLC X (X-box)domain. Both the X-box domain and the Y-box domain are important for catalytic activity in PLC proteins. X-box domains are conserved from prokaryotes to mammals. The gene encoding PLC-XD2 maps to human chromosome 3, which is made up of about 214 million bases encoding over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. There are two isoforms of PLC-XD2 that are produced as a result of alternative splicing events.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLCXD2
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLCXD2
Gene Accession No. Q0VAA5
Gene Alias AW494639; EG433022; phosphatidylinositol specific phospholipase C X domain containing 2; phosphatidylinositol-specific phospholipase C, X domain containing 2; PI-PLC X domain-containing protein 2; PLCXD2; RGD1563504
Gene Symbols PLCXD2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLCXD2. Recombinant protein control fragment (Product #RP-100580).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 257068
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.