Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLCXD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA576231
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
PLC-XD2 (PI-PLC X domain-containing protein 2) is a 305 amino acid protein that contains a domain that is present in many PLC isozymes, the PI-PLC X (X-box)domain. Both the X-box domain and the Y-box domain are important for catalytic activity in PLC proteins. X-box domains are conserved from prokaryotes to mammals. The gene encoding PLC-XD2 maps to human chromosome 3, which is made up of about 214 million bases encoding over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. There are two isoforms of PLC-XD2 that are produced as a result of alternative splicing events.
Specifications
PLCXD2 | |
Polyclonal | |
Unconjugated | |
PLCXD2 | |
AW494639; EG433022; phosphatidylinositol specific phospholipase C X domain containing 2; phosphatidylinositol-specific phospholipase C, X domain containing 2; PI-PLC X domain-containing protein 2; PLCXD2; RGD1563504 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
257068, 363781, 433022 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
B2RXA1, Q0VAA5 | |
PLCXD2 | |
Recombinant fusion protein of human PLCXD2. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction