Learn More
Invitrogen™ PLCXD2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121120
Description
Positive test controls include: NIH/3T3, SH-SY5Y, U-251MG, Mouse liver, Mouse brain, Mouse pancreas, Mouse eye, Rat liver. Immunogen sequence: MLAVRKARRK LRMGTICSPN PSGTKTSSEV CNADWMASLP PHLHNLPLSN LAIPGSHDSF SYWVDEKSPV GPDQTQAIKR LARISLVKKL MKKWSVTQNL TFREQLEAGI RYFDLRVSSK PGDADQEIYF IHGLFGIKVW DGLMEIDSFL TQHPQEIIFL DFNHFYAMDE THHKCLVLRI QEAFGNKLCP ACSVESLTLR TLWEKNCQVL IFYHCPFYKQ YPFLWPGKKI PAPWANTTSV RKLILFLETT LSERASRGSF HVSQAILTPR VKTIARGLVG GLKNTLVHSN RWNSHGPSLL SQERS.
PLC-XD2 (PI-PLC X domain-containing protein 2) is a 305 amino acid protein that contains a domain that is present in many PLC isozymes, the PI-PLC X (X-box)domain. Both the X-box domain and the Y-box domain are important for catalytic activity in PLC proteins. X-box domains are conserved from prokaryotes to mammals. The gene encoding PLC-XD2 maps to human chromosome 3, which is made up of about 214 million bases encoding over 1,100 genes, including a chemokine receptor (CKR) gene cluster and a variety of human cancer-related gene loci. There are two isoforms of PLC-XD2 that are produced as a result of alternative splicing events.
Specifications
PLCXD2 | |
Polyclonal | |
Unconjugated | |
PLCXD2 | |
AW494639; EG433022; phosphatidylinositol specific phospholipase C X domain containing 2; phosphatidylinositol-specific phospholipase C, X domain containing 2; PI-PLC X domain-containing protein 2; PLCXD2; RGD1563504 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
257068, 363781, 433022 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
3.23 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
B2RXA1, Q0VAA5 | |
PLCXD2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-305 of human PLCXD2 (NP_001172035.1). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.