Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLCXD3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA561351
Description
Immunogen sequence: QFLQASITER RKKGSFFISQ VVLTPKASTV VKGVASGLRE TITERALPAM MQWVRTQKPG ESGINIVTAD FVELGDFIST VIKLNYVFDE GEANT Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 99%.
PLCXD3 gene ontology annotations related to this gene include lipid metabolic process.
Specifications
PLCXD3 | |
Polyclonal | |
Unconjugated | |
PLCXD3 | |
B130016O10Rik; phosphatidylinositol specific phospholipase C X domain containing 3; phosphatidylinositol-specific phospholipase C, X domain containing 3; PI-PLC X domain-containing protein 3; PLCXD3; RGD1308448 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
345557 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q63HM9 | |
PLCXD3 | |
Recombinant protein corresponding to Human PLCXD3. Recombinant protein control fragment (Product #RP-96968). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction