Learn More
PLD1/Phospholipase D1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305241A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Phosphatidylcholine (PC)-specific phospholipases D (PLDs; EC 3.1.4.4) catalyze the hydrolysis of PC to produce phosphatidic acid and choline. A range of agonists acting through G protein-coupled receptors and receptor tyrosine kinases stimulate this hydrolysis. PC-specific PLD activity has been implicated in numerous cellular pathways, including signal transduction, membrane trafficking, and the regulation of mitosis (Hammond et al., 1995).Specifications
PLD1/Phospholipase D1 | |
Polyclonal | |
Unconjugated | |
PLD1 | |
PLD 1; choline phosphatase 1; hPLD1; phosphatidylcholine-hydrolyzing phospholipase D1; phospholipase D1; phospholipase D1, phophatidylcholine-specific | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5337 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q13393 | |
PLD1 | |
Between 525 and 575 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.