Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583606
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583606 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583606 Supplier Invitrogen™ Supplier No. PA583606
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: NEPRVNTSAL QKIAADMSNI IENLDTRELH FEGEEVDYDV SPSDPKIQEV YIPFSAIYNT QGFKEPNIQT YLSGCPIKAQ VLEVERFTST TRVPSINLYT IELTHGEFKW QVKRKFKHFQ EFHR Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 85%.

Phosphatidylcholine (PC)-specific phospholipases D (PLDs; EC 3.1.4.4) catalyze the hydrolysis of PC to produce phosphatidic acid and choline. A range of agonists acting through G protein-coupled receptors and receptor tyrosine kinases stimulate this hydrolysis. PC-specific PLD activity has been implicated in numerous cellular pathways, including signal transduction, membrane trafficking, and the regulation of mitosis (Hammond et al., 1995).
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLD1
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PLD1
Gene Accession No. Q13393
Gene Alias AA536939; C85393; choline phosphatase 1; hPLD1; mPLD1; phophatidylcholine-specific; Phoshpolipase D gene 1 probably with two splicing variants A and B; phosphatidylcholine-hydrolyzing phospholipase D1; phospholipase D gene 1, probably with two splicing variants A and B; phospholipase D1; phospholipase D1, phosphatidylcholine-specific; PLD 1; Pld1; Pld1a; Pld1b; Plda; Pldb; rPLD1
Gene Symbols PLD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PLD1. Recombinant protein control fragment (Product #RP-90700).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5337
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.