Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLD2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587702
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587702 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587702 Supplier Invitrogen™ Supplier No. PA587702
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MTATPESLFP TGDELDSSQL QMESDEVDTL KEGEDPADRM HPFLAIYELQ SLKVHPLVFA PGVPVTAQVV GTERYTSGSK VGTCTLYSVR LTHGDFSWTT KKKYRHFQEL HRDLLRHKVL MSLLPLARFA VAYSPARDAG NREMPSLPRA GPEGSTRHAA SKQKYLENYL NRLLTMSFYR; Positive Samples: B-cell, HeLa, U-87MG, BT-474, A375; Cellular Location: Membrane, Peripheral membrane protein.

PLD2 is a peripheral membrane protein belonging to phospholipase D family with a PH domain, two PLD phosphodiesterase domains and a PX (phox homology) domain that plays a regulatory rather than catalytic role. PLD2 catalyzes the hydrolysis of phosphatidylcholine (PC) to produce phosphatidic acid and choline. PLD2 may function in regulated secretion, signal-induced cytoskeletal regulation and/or endocytosis, transcriptional regulation, and cell cycle control. It is usually stimulated by phosphatidylinositol 4,5-bisphosphate (PIP2) and activated by the ADP-ribosylation factor-1 (ARF-1). PLD2 is activated by agonist stimulation of both tyrosine kinase and G protein-coupled receptors and is also known to interact with EGFR and PIP5K1A. PLD2, localized to plasma membrane caveolae, is ubiquitously expressed in most tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLD2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 3.55 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Pld2
Gene Accession No. P70498, P97813
Gene Alias choline phosphatase 2; hPLD2; phosphatidylcholine-hydrolyzing phospholipase D2; phospholipase D gene 2; phospholipase D2; PLD 2; PLD1C; PLD2; Pldc; rPLD2
Gene Symbols Pld2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-180 of human PLD2 (NP_0026543).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18806, 25097
Target Species Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.