Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA531959
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA531959 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA531959 Supplier Invitrogen™ Supplier No. PA531959
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A549. Predicted reactivity: Mouse (92%), Rat (92%), Pig (95%), Rhesus Monkey (98%), Bovine (96%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

PLD3 is a ingle-pass type II membrane protein. It belongs to the phospholipase D family. PLD3 contains 2 PLD phosphodiesterase domains. The exact function of PLD3 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLD3
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene PLD3
Gene Accession No. Q8IV08
Gene Alias 5'-3' exonuclease PLD3; phospholipase D3; AD19; Choline phosphatase 3; hindIII K4L homolog; HUK4; HU-K4; phosphatidylcholine-hydrolyzing phospholipase D3; phospholipase D family member 3; phospholipase D family, member 3; phospholipase D3; PLD 3; Pld3; Sam9; Sam-9; Schwannoma-associated protein 9; Unknown (protein for MGC:126910)
Gene Symbols PLD3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human PLD3. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23646
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.