Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542640
Description
Peptide sequence: WVLLVLILAV VGFGALMTQL FLWEYGDLHL FGPNQRPAPC YDPCEAVLVE Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
PLD3 is a ingle-pass type II membrane protein. It belongs to the phospholipase D family. PLD3 contains 2 PLD phosphodiesterase domains. The exact function of PLD3 remains unknown.
Specifications
| PLD3 | |
| Polyclonal | |
| Unconjugated | |
| PLD3 | |
| 5'-3' exonuclease PLD3; phospholipase D3; AD19; Choline phosphatase 3; hindIII K4L homolog; HUK4; HU-K4; phosphatidylcholine-hydrolyzing phospholipase D3; phospholipase D family member 3; phospholipase D family, member 3; phospholipase D3; PLD 3; Pld3; Sam9; Sam-9; Schwannoma-associated protein 9; Unknown (protein for MGC:126910) | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 23646 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot, Surface Plasmon Resonance | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IV08 | |
| PLD3 | |
| Synthetic peptide directed towards the N-terminal of human PLD3 (aa 38-87). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction