Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104016
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104016 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104016 Supplier Invitrogen™ Supplier No. PA5104016
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total PLD3.

PLD3 is a ingle-pass type II membrane protein. It belongs to the phospholipase D family. PLD3 contains 2 PLD phosphodiesterase domains. The exact function of PLD3 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLD3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLD3
Gene Accession No. O35405, Q5FVH2, Q8IV08
Gene Alias 5'-3' exonuclease PLD3; phospholipase D3; AD19; Choline phosphatase 3; hindIII K4L homolog; HUK4; HU-K4; phosphatidylcholine-hydrolyzing phospholipase D3; phospholipase D family member 3; phospholipase D family, member 3; phospholipase D3; PLD 3; Pld3; Sam9; Sam-9; Schwannoma-associated protein 9; Unknown (protein for MGC:126910)
Gene Symbols PLD3
Host Species Rabbit
Immunogen A synthesized peptide derived from human PLD3(Accession Q8IV08), corresponding to amino acid residues G452-L490.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18807, 23646, 361527
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.