Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLD3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120638
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120638 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120638 Supplier Invitrogen™ Supplier No. PA5120638
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Mouse testis, Mouse lung, Mouse brain. Immunogen sequence: MEICLNGTPA LAYLASAPPP LCPSGRTPDL KALLNVVDNA RSFIYVAVMN YLPTLEFSHP HRFWPAIDDG LRRATYERGV KVRLLISCWG HSEPSMRAFL LSLAALRDNH THSDIQVKLF VVPADEAQAR IPYARVNHNK YMVTERATYI GTSNWSGNYF TETAGTSLLV TQNGRGGLRS QLEAIFLRDW DSPYSHDLDT SADSVGNACR LL.

PLD3 is a ingle-pass type II membrane protein. It belongs to the phospholipase D family. PLD3 contains 2 PLD phosphodiesterase domains. The exact function of PLD3 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLD3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.12 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene PLD3
Gene Accession No. O35405
Gene Alias 5'-3' exonuclease PLD3; phospholipase D3; AD19; Choline phosphatase 3; hindIII K4L homolog; HUK4; HU-K4; phosphatidylcholine-hydrolyzing phospholipase D3; phospholipase D family member 3; phospholipase D family, member 3; phospholipase D3; PLD 3; Pld3; Sam9; Sam-9; Schwannoma-associated protein 9; Unknown (protein for MGC:126910)
Gene Symbols PLD3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 279-490 of human PLD3 (NP_001026866).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18807
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.