Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLD3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | PLD3 |
---|---|
Concentration | 0.27 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1511
![]() ![]() |
Proteintech Group Inc
173271AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
PLD3 is a ingle-pass type II membrane protein. It belongs to the phospholipase D family. PLD3 contains 2 PLD phosphodiesterase domains. The exact function of PLD3 remains unknown.Specifications
PLD3 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O35405, Q5FVH2, Q8IV08 | |
18807, 23646, 361527 | |
PLD3 Fusion Protein Ag11306 | |
Primary | |
-20°C | |
PLD3 |
0.27 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Choline phosphatase 3, HindIII K4L homolog, HU K4, Phospholipase D3, PLD 3, PLD3 | |
Pld3 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title