Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ PLD4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA550089
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA550089 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA550089 Supplier Invitrogen™ Supplier No. PA550089
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total PLD4 protein.

PLD4 (Phospholipase D Family Member 4) is a Protein Coding gene. Among its related pathways are Innate Immune System and Inositol phosphate metabolism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PLD4
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene PLD4
Gene Accession No. Q8BG07, Q96BZ4
Gene Alias AI132321; C14orf175; Choline phosphatase 4; phosphatidylcholine-hydrolyzing phospholipase D4; phospholipase D family member 4; phospholipase D family, member 4; phospholipase D4; PLD 4; PLD4; thss; UNQ2488/PRO5775
Gene Symbols PLD4
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human PLD4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104759, 122618, 362792
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.