Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLD4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA562425
Description
Immunogen sequence: AQDLEKTFQT YWVLGVPKAV LPKTWPQNFS SHFNRFQPFH GLFDGVPTTA YFSASPPALC PQGRTRDLEA LLAVMGSAQE FIYASVMEYF P Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 81%.
PLD4 (Phospholipase D Family Member 4) is a Protein Coding gene. Among its related pathways are Innate Immune System and Inositol phosphate metabolism.
Specifications
PLD4 | |
Polyclonal | |
Unconjugated | |
PLD4 | |
AI132321; C14orf175; Choline phosphatase 4; phosphatidylcholine-hydrolyzing phospholipase D4; phospholipase D family member 4; phospholipase D family, member 4; phospholipase D4; PLD 4; PLD4; thss; UNQ2488/PRO5775 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
122618 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q96BZ4 | |
PLD4 | |
Recombinant protein corresponding to Human PLD4. Recombinant protein control fragment (Product #RP-101451). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction