Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ PLD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113180
Description
PLD6 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
PLD6 is a cardiolipin hydrolase present at the mitochondrial outer membrane required both for mitochondrial fusion and piRNA metabolic process. PLD6 acts by catalyzing the hydrolysis of cardiolipin (diphosphatidylglycerol) to form phosphatidate (phosphatidic acid or PA) at the mitochondrial outer membrane surface, promoting mitochondrial fusion. The production of phosphatidate also regulates the piRNA metabolic process by promoting recruitment and/or activation of components of the meiotic nuage, also named P granule, a critical step for primary biogenesis of piRNAs. It is required during spermatogenesis to repress transposable elements and prevent their mobilization via its role in the piRNA metabolic process, which mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and govern the methylation and subsequent repression of transposons.
Specifications
| PLD6 | |
| Polyclonal | |
| Unconjugated | |
| PLD6 | |
| 4933433K01Rik; choline phosphatase 6; Gm10; Mitochondrial cardiolipin hydrolase; mitochondrial phospholipase; mitoPLD; mZuc; novel Phospholipase D Active site motif-containing protein; phosphatidylcholine-hydrolyzing phospholipase D6; phospholipase D family member 6; phospholipase D family, member 6; phospholipase D6; PLD 6; PLD6; Protein zucchini homolog; Zuc | |
| Rabbit | |
| Protein G | |
| RUO | |
| 194908, 201164 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q5SWZ9, Q8N2A8 | |
| PLD6 | |
| Recombinant Human Mitochondrial cardiolipin hydrolase protein (1-252AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction