Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Pleckstrin-2 Antibody (OTI5D3), Alexa Fluor™ 647, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273467AF647
Description
Pleckstrin-2 Monoclonal specifically detects Pleckstrin-2 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
Pleckstrin-2 | |
Monoclonal | |
Alexa Fluor 647 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PLEK2 | |
Recombinant protein fragment corresponding to amino acids 132-353 of human Pleckstrin-2 (NP_057529) produced in E.coli. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot | |
OTI5D3 | |
Western Blot | |
pleckstrin 2, pleckstrin 2 homolog, pleckstrin-2 | |
Mouse | |
Affinity Purified | |
Signal Transduction | |
26499.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction