Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Pleckstrin-2 Antibody (OTI5D3), DyLight 755, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

Supplier:  Novus Biologicals NBP273467IR

Catalog No. NBP273467IR


Only null left
Add to Cart

Description

Description

Pleckstrin-2 Monoclonal specifically detects Pleckstrin-2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

Pleckstrin-2
Monoclonal
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
PLEK2
Recombinant protein fragment corresponding to amino acids 132-353 of human Pleckstrin-2 (NP_057529) produced in E.coli.
0.1 mL
Primary
Human, Mouse, Rat
IgG1
Western Blot
OTI5D3
Western Blot
pleckstrin 2, pleckstrin 2 homolog, pleckstrin-2
Mouse
Affinity Purified
Signal Transduction
26499.0
Store at 4°C in the dark.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.