Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pleckstrin 2 Monoclonal Antibody (OTI5D3)
GREENER_CHOICE

Catalog No. PIMA526551
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA526551 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA526551 Supplier Invitrogen™ Supplier No. MA526551
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Pleckstrin 2 Monoclonal Antibody for Western Blot

Pleckstrin 2 may help orchestrate cytoskeletal arrangement and contribute to lamellipodia formation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pleckstrin 2
Applications Western Blot
Classification Monoclonal
Clone OTI5D3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Plek2
Gene Accession No. Q9NYT0
Gene Alias pleckstrin 2; pleckstrin 2 homolog; pleckstrin-2; PLEK2
Gene Symbols Plek2
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 132-353 of PLEK2 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26499
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.