Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pleckstrin Monoclonal Antibody (2D8)
GREENER_CHOICE

Catalog No. PIMA519629
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA519629 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA519629 Supplier Invitrogen™ Supplier No. MA519629
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: PETIDLGALY LSMKDTEKGI KELNLEKDKK IFNHCFTGNC VIDWLVSNQS VRNRQEGLMI ASSLLNEGYL QPAGDMSKSA VDGTAENPFL DNPDAFYYFP DSGFFCEENS.

Major protein kinase C substrate of platelets.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pleckstrin
Applications ELISA, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 2D8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene PLEK
Gene Accession No. P08567
Gene Alias 2010300B13Rik; FLJ27168; P47; platelet 47 kDa protein; Pleckstrin; Plek
Gene Symbols PLEK
Host Species Mouse
Immunogen PLEK (AAH18549, 121 a.a. ~ 230 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5341
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.