Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Pleckstrin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521753
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521753 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521753 Supplier Invitrogen™ Supplier No. PA521753
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: Raji. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Major protein kinase C substrate of platelets.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pleckstrin
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.61 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene PLEK
Gene Accession No. P08567
Gene Alias 2010300B13Rik; FLJ27168; P47; platelet 47 kDa protein; Pleckstrin; Plek
Gene Symbols PLEK
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Pleckstrin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5341
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.