Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Pleckstrin Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA589170
Description
Immunogen sequence: YLQPAGDMSK SAVDGTAENP FLDNPDAFYY FPDSGFFCEE NSSDDDVILK EEFRGVIIKQ GCLLKQGHRR KNWKVRKFIL REDPAYLHYY DPAGAEDPLG AIHLRGCVVT SVESNSNGRK SEEENLFEII TADEVHYFLQ AATPKERTEW IRAIQMASRT GK; Positive Samples: THP-1, U-937, Raji.
Major protein kinase C substrate of platelets.
Specifications
Pleckstrin | |
Polyclonal | |
Unconjugated | |
PLEK | |
2010300B13Rik; FLJ27168; P47; platelet 47 kDa protein; Pleckstrin; Plek | |
Rabbit | |
Affinity Chromatography | |
RUO | |
5341 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
5.58 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08567 | |
PLEK | |
Recombinant fusion protein containing a sequence corresponding to amino acids 189-350 of human PLEK (NP_0026552). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction