Learn More
Invitrogen™ Pleckstrin Recombinant Rabbit Monoclonal Antibody (4W2K3)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA555527

Description
Immunogen Sequence: MEPKRIREGY LVKKGSVFNT WKPMWVVLLE DGIEFYKKKS DNSPKGMIPL KGSTLTSPCQ DFGKRMFVFK ITTTKQQDHF FQAAFLEERD AWVRDIKKAI.
Major protein kinase C substrate of platelets.
Specifications
Pleckstrin | |
Recombinant Monoclonal | |
0.2 mg/mL | |
PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P08567, Q9JHK5 | |
PLEK | |
A synthetic peptide corresponding to a sequence within amino acids 1-100 of human Pleckstrin. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
ELISA, Western Blot | |
4W2K3 | |
Unconjugated | |
PLEK | |
2010300B13Rik; FLJ27168; P47; platelet 47 kDa protein; Pleckstrin; Plek | |
Rabbit | |
Protein A | |
RUO | |
5341, 56193 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.