Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plectin Antibody (7A8), Novus Biologicals™
SDP

Catalog No. NB310285 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 ml
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310285 0.025 ml
NB12011220 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB310285 Supplier Novus Biologicals Supplier No. NB120112200.025ML
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 1 publication

Plectin Monoclonal specifically detects Plectin in Human, Mouse, Rat, Bovine, Mammal samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen.

Specifications

Antigen Plectin
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry
Classification Monoclonal
Clone 7A8
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Gene Alias EBS1, HD1, Hemidesmosomal protein 1, LGMD2Q, PCNplectin 1, intermediate filament binding protein, 500kD, PLEC1b, PLEC1EBSO, plectin, plectin 1, intermediate filament binding protein 500kDa, plectin-1, PLTNepidermolysis bullosa simplex 1 (Ogna)
Gene Symbols PLEC
Host Species Mouse
Immunogen Plectin purified from rat glioma C6 cells.
Purification Method Unpurified
Quantity 0.025 ml
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5339
Test Specificity The inhibits the interaction of plectin with vimentin and lamin B.
Target Species Mammalia, Human, Rat, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.