Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plectin Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501567893
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-7893 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-7893 Supplier Bethyl Laboratories, Inc Supplier No. A304506AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Rat

Plectin is a prominent member of an important family of structurally and in part functionally related proteins, termed plakins or cytolinkers, that are capable of interlinking different elements of the cytoskeleton. Plakins, with their multi-domain structure and enormous size, not only play crucial roles in maintaining cell and tissue integrity and orchestrating dynamic changes in cytoarchitecture and cell shape, but also serve as scaffolding platforms for the assembly, positioning, and regulation of signaling complexes. Plectin is expressed as several protein isoforms in a wide range of cell types and tissues from a single gene located on chromosome 8 in humans. The expression of specific plectin isoforms was found to be dependent on cell type (tissue) and stage of development and it appears that each cell type (tissue) contains a unique set (proportion and composition) of plectin isoforms which carry out distinct and specific functions.

Specifications

Antigen Plectin
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PLEC
Gene Accession No. Q15149, Q9QXS1
Gene Alias EBS1, EBSMD, EBSND, EBSO, EBSOG, EBSPA, epidermolysis bullosa simplex 1 (Ogna), HD1, hemidesmosomal protein 1, LGMD2Q, PCN, PLEC1, PLEC1b, plectin 1, intermediate filament binding protein 500kDa, plectin 1, intermediate filament binding protein, 500kD, Plectin-1, PLTN
Gene Symbols PLEC
Host Species Rabbit
Immunogen Between 4534 and 4684
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18810, 5339
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.