Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Plectin Recombinant Rabbit Monoclonal Antibody (9O0B3), Invitrogen™

Catalog No. PIMA542733
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA542733 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIMA542733 Supplier Thermo Scientific Supplier No. MA542733
This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: RTQLASWSDP TEETGPVAGI LDTETLEKVS ITEAMHRNLV DNITGQRLLE AQACTGGIID PSTGERFPVT DAVNKGLVDK IMVDRINLAQ KAFCGFEDPR TKTKMSAAQA LKKGWLYYEA GQRFLEVQYL TGGLIEPDTP GRVPLDEALQ RGTVDARTAQ KLRDVGAYSK YLTCPKTKLK ISYKDALDRS MVEEGTGLRL LEAAAQSTK

Plectin is a prominent member of an important family of structurally and in part functionally related proteins, termed plakins or cytolinkers, that are capable of interlinking different elements of the cytoskeleton. Plakins, with their multi-domain structure and enormous size, not only play crucial roles in maintaining cell and tissue integrity and orchestrating dynamic changes in cytoarchitecture and cell shape, but also serve as scaffolding platforms for the assembly, positioning, and regulation of signaling complexes. Plectin is expressed as several protein isoforms in a wide range of cell types and tissues from a single gene located on chromosome 8 in humans. The expression of specific plectin isoforms was found to be dependent on cell type (tissue) and stage of development and it appears that each cell type (tissue) contains a unique set (proportion and composition) of plectin isoforms which carry out distinct and specific functions.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Plectin
Applications Immunohistochemistry (Paraffin)
Classification Recombinant Monoclonal
Clone 9O0B3
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene Plec
Gene Accession No. P30427, Q15149
Gene Alias AA591047; AU042537; EBS1; EBSMD; EBSND; EBSO; EBSOG; EBSPA; HD1; Hemidesmosomal protein 1; LGMD2Q; PCN; Plec; PLEC1; PLEC1b; Plectin; plectin 1; plectin 1, intermediate filament binding protein 500kDa; plectin-1; Plectin-6; Pltn
Gene Symbols Plec
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 4401-4609 of human Plectin (NP_9587821)
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5339, 64204
Target Species Human, Rat
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.