Learn More
Invitrogen™ Pleiotrophin Polyclonal Antibody

Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:8,000.
Specifications
Specifications
| Antigen | Pleiotrophin |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | PTN |
| Gene Accession No. | P21246, P63089 |
| Gene Alias | HARP; HBBM; HBBN; HB-GAM; HBGF8; HBGF-8; HBNF; HBNF1; HBNF-1; heparin affin regulatory peptide; heparin affin regulatory protein; heparin binding factor; heparin-binding brain mitogen; Heparin-binding growth factor 8; heparin-binding growth-associated molecule; heparin-binding neurite outgrowth promoting factor; Heparin-binding neurite outgrowth-promoting factor; Heparin-binding neurite outgrowth-promoting factor 1; heparin-binding neurite promoting factor; heparin-binding neurotorphic factor; heparin-binding neutrophic factor; NEGF1; OSF; Osf1; OSF-1; osteoblastic cell factor; Osteoblast-specific factor 1; pleiotrophin; pleiotrophin (heparin binding growth factor 8, neurite growth-promoting factor 1); Pleiotrophin (Heparine binding factor, Hbnf, in the mouse); PTN |
| Gene Symbols | PTN |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.