Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PLEK Mouse anti-Human, Porcine, Clone: 1D5C2, Proteintech
SDP

Catalog No. 501736858 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6858 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6858 Supplier Proteintech Group Inc Supplier No. 664311IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Major protein kinase C substrate of platelets.

Specifications

Antigen PLEK
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 1D5C2
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene PLEK
Gene Accession No. P08567
Gene Alias P47, Platelet 47 kDa protein, pleckstrin, PLEK
Gene Symbols PLEK
Host Species Mouse
Immunogen PLEK Fusion Protein Ag3187
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100516257, 5341
Target Species Human, Pig
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.