Learn More
PLEKHA1/TAPP1 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A305233AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
Binds specifically to phosphatidylinositol 3,4-diphosphate (PtdIns3,4P2), but not to other phosphoinositides. May recruit other proteins to the plasma membrane.Specifications
PLEKHA1/TAPP1 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q9HB21 | |
PLEKHA1 | |
Between 354 and 404 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
PLEKHA1 | |
PLEKHA1, family A (phosphoinositide bindingspecific) member 1, PH domain-containing family A member 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
59338 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.