Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHA2 Antibody, DyLight 405, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306069V
Description
PLEKHA2 Polyclonal specifically detects PLEKHA2 in Human samples. It is validated for Western Blot.Specifications
PLEKHA2 | |
Polyclonal | |
Western Blot | |
FLJ25921, PH domain-containing family A member 2, pleckstrin homology domain containing, family A (phosphoinositide binding specific) member 2, pleckstrin homology domain-containing family A member 2, pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2, tandem PH Domain containing protein-2, tandem PH domain-containing protein 2, TAPP2, TAPP-2 | |
Produced in rabbits immunized with E. coli-derived Human PLEKHA2 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
DyLight 405 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
59339 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction