Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHA3 Antibody (OTI5D11), Alexa Fluor™ 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273470AF405
Description
PLEKHA3 Monoclonal specifically detects PLEKHA3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
PLEKHA3 | |
Monoclonal | |
Alexa Fluor 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
PLEKHA3 | |
Full length recombinant protein of human PLEKHA3 (NP_061964) produced in HEK293T cells. | |
0.1 mL | |
65977.0 | |
Store at 4°C in the dark. |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI5D11 | |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
FAPP-1, FAPP1PH domain-containing family A member 3, FLJ20067, four-phosphate-adaptor protein 1, Phosphatidylinositol-four-phosphate adapter protein 1, Phosphoinositol 4-phosphate adapter protein 1, pleckstrin homology domain containing, family A (phosphoinositide bindingspecific) member 3, pleckstrin homology domain-containing family A member 3, pleckstrin homology domain-containing, family A (phosphoinositide bindingspecific) member 3 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction