Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PLEKHA3 Monoclonal Antibody (OTI6E4), TrueMAB™, OriGene

Mouse Monoclonal Antibody
Supplier: OriGene TA502876

Description
PLEKHA3 (Pleckstrin Homology Domain Containing A3) is a Protein Coding gene. Among its related pathways are Metabolism and Cholesterol and Sphingolipids transport / Transport from Golgi and ER to the apical membrane (normal and CF). Gene Ontology (GO) annotations related to this gene include phospholipid binding and 1-phosphatidylinositol binding. An important paralog of this gene is PLEKHA8.Specifications
PLEKHA3 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
Q9HB20 | |
PLEKHA3 | |
Full length human recombit protein of human PLEKHA3 produced in HEK293T cell. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI6E4 | |
Unconjugated | |
PLEKHA3 | |
FAPP1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
65977 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction